Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099779.1 | NP_001093249.1 | 2 | 134 | |
NM_013631.2 | NP_038659.2 | 2 | 260 | ||
GenBank mRNA | AK149508.1 | - | 1 | 135 | |
AK149524.1 | - | 1 | 133 | ||
BC152327.1 | - | 2 | 244 | 40141144 | |
BC152542.1 | - | 2 | 244 | ||
BY107231.1 | - | 2 | 228 | ||
BY781095.1 | - | 2 | 260 | ||
D63764.1 | - | 2 | 260 | ||
S79731.1 | - | 2 | 260 |