Gene Transcripts
- Gene Symbol
- Sars
- Entrez Gene ID
- 20226
- Gene Name
- seryl-aminoacyl-tRNA synthetase
- Gene Aliases
- Sars1, Strs, serRS
- Chromosome Location
- Chr. 3: 108424864 - 108445259 on Build GRCm38
- UniGene ID
- Mm.28688
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204979.1 |
NP_001191908.1
|
3, 4 |
423 |
|
NM_011319.3 |
NP_035449.1
|
3, 4 |
423 |
|
GenBank mRNA |
AK028358.1 |
- |
3, 4 |
355 |
|
AK075827.1 |
- |
3, 4 |
354 |
|
AK077309.1 |
- |
3, 4 |
305 |
|
AK082793.1 |
- |
3, 4 |
334 |
|
AK152150.1 |
- |
3, 4 |
381 |
|
AK153158.1 |
- |
3, 4 |
373 |
|
AK217272.1 |
- |
1, 2 |
48 |
|
BC008612.1 |
- |
3, 4 |
312 |
3257101 |
BC031932.1 |
- |
3, 4 |
312 |
|
View Full Product Details