Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130456.1 | NP_001123928.1 | 9 | 906 | |
NM_013662.2 | NP_038690.1 | 9 | 1017 | ||
GenBank mRNA | AF036585.1 | - | 8 | 773 | |
AK139358.1 | - | 9 | 826 | ||
AK143229.1 | - | 9 | 907 | ||
AK166077.1 | - | 9 | 1009 | ||
BC139415.1 | - | 8 | 772 | 8862437 |