Gene Transcripts

Gene Symbol
Prrg3
Entrez Gene ID
208748
Gene Name
proline rich Gla (G-carboxyglutamic acid) 3 (transmembrane)
Gene Aliases
Gm368
Chromosome Location
Chr. X: 71962156 - 71972722 on Build GRCm38
UniGene ID
Mm.25626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC060135.1 - 4 599
BC137616.1 - 4 728 8860635
BC137619.1 - 4 728 8860638

View Full Product Details