Gene Transcripts

Gene Symbol
Tsga10
Entrez Gene ID
211484
Gene Name
testis specific 10
Gene Aliases
4933432N21Rik, AI843383, Gm217, Mtsga10
Chromosome Location
Chr. 1: 37728144 - 37866739 on Build GRCm38
UniGene ID
Mm.332756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290720.1 NP_001277649.1 15 2044
NM_001290721.1 NP_001277650.1 14 1851
NM_207228.3 NP_997111.1 15 1911
GenBank mRNA AF530050.3 - 14 1856
AK030254.1 - 15 2044
AY618264.1 - 14 1856
BC053407.1 - 13 1679
BC066782.1 - 15 1908 6705127
CK789086.1 - 4 420

View Full Product Details