Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040435.2 | NP_001035525.1 | 9 | 1834 | |
GenBank mRNA | AF093542.1 | - | 10 | 1068 | |
AF156934.1 | - | 9 | 1388 | ||
AF203445.1 | - | 9 | 1335 | ||
AF203446.1 | - | 9 | 723 | ||
AF247674.1 | - | 10 | 1119 | ||
AK076282.1 | - | 1861 | |||
AK145556.1 | - | 9 | 1395 | ||
AK160291.1 | - | 9 | 1397 | ||
AK183147.1 | - | 1 | 48 | ||
AK185221.1 | - | 3 | 160 | ||
BC003252.1 | - | 9 | 1391 | 3500413 | |
BC055071.1 | - | 3810 | |||
BC066184.1 | - | 9 | 1370 | 30457053 | |
BC068314.1 | - | 9 | 1370 | 30461783 |