Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178045.4 | NP_835146.3 | 7 | 757 | |
GenBank mRNA | AK036528.1 | - | 7 | 721 | |
AK089460.1 | - | 7 | 715 | ||
AK139284.1 | - | 7 | 704 | ||
AK151561.1 | - | 7 | 722 | ||
AK152639.1 | - | 7 | 722 | ||
AK160584.1 | - | 7 | 731 | ||
AK164357.1 | - | 7 | 720 | ||
AK170955.1 | - | 7 | 706 | ||
AK171198.1 | - | 7 | 735 | ||
BC060709.1 | - | 7 | 712 | 6409692 |