Gene Transcripts

Gene Symbol
Lrrc1
Entrez Gene ID
214345
Gene Name
leucine rich repeat containing 1
Gene Aliases
A430093J20Rik, AU016091, D030059I21, mKIAA4018
Chromosome Location
Chr. 9: 77430823 - 77544852 on Build GRCm38
UniGene ID
Mm.28534
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146048.1 NP_001139520.1 14 1745
NM_172528.3 NP_766116.3 14 1393
GenBank mRNA AK082721.1 - 14 1394
AK147075.1 - 14 1745
AK220166.1 - 14 1763
BC022613.1 - 6 687
BC046591.1 - 14 1732
BC087542.1 - 14 1592 4934980

View Full Product Details