Gene Transcripts

Gene Symbol
Senp6
Entrez Gene ID
215351
Gene Name
SUMO/sentrin specific peptidase 6
Gene Aliases
2810017C20Rik, E130319N12Rik, Susp1, mKIAA0797
Chromosome Location
Chr. 9: 80066516 - 80144782 on Build GRCm38
UniGene ID
Mm.28232
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146003.2 NP_666115.2 7 1269
GenBank mRNA AK048485.1 - 2 718
AK083377.1 - 7 858
AK129219.1 - 3 796
BC052718.1 - 7 1036
BC061480.1 - 7 1093 6509771
BC151128.1 - 8 700 9088041
BC172171.1 - 8 700 9054841

View Full Product Details