Gene Transcripts

Gene Symbol
Slc16a5
Entrez Gene ID
217316
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 5
Gene Aliases
A130015N09Rik, MCT 5, MCT 6, MCT5
Chromosome Location
Chr. 11: 115462250 - 115474398 on Build GRCm38
UniGene ID
Mm.25773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080934.1 NP_001074403.1 1 240
GenBank mRNA AK079757.1 - 1 242
BC172170.1 - 1 152

View Full Product Details