Gene Transcripts

Gene Symbol
Nploc4
Entrez Gene ID
217365
Gene Name
nuclear protein localization 4 homolog (S. cerevisiae)
Gene Aliases
AK129375, Npl4, mKIAA1499
Chromosome Location
Chr. 11: 120379798 - 120437700 on Build GRCm38
UniGene ID
Mm.309520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195023.1 NP_001181952.1 3 324
NM_199469.2 NP_955763.1 3 324
GenBank mRNA AK129375.1 - 3 299
AK141998.1 - 3 323
AK149823.1 - 3 296
AK149912.1 - 3 323
BC065156.1 - 3 292 5706754

View Full Product Details