Gene Transcripts

Gene Symbol
Lrrc45
Entrez Gene ID
217366
Gene Name
leucine rich repeat containing 45
Gene Aliases
BC023296
Chromosome Location
Chr. 11: 120713953 - 120721128 on Build GRCm38
UniGene ID
Mm.347545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153545.2 NP_705773.2 12 1350
GenBank mRNA AK081566.1 - 12 1349
AK153361.1 - 12 1351
BC023296.1 - 12 1330 3662961

View Full Product Details