Gene Transcripts

Gene Symbol
Dnajc27
Entrez Gene ID
217378
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 27
Gene Aliases
AI639580, C330021A05Rik, Rabj, Rbj
Chromosome Location
Chr. 12: 4082568 - 4118013 on Build GRCm38
UniGene ID
Mm.271656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153082.4 NP_694722.2 6 839
GenBank mRNA AK166860.1 - 1 1324
AY035893.1 - 6 626
BC042643.1 - 6 754 5357595
BC051140.1 - 6 801 6595101
CB247441.1 - 5 553

View Full Product Details