Gene Transcripts
- Gene Symbol
- Mtmr6
- Entrez Gene ID
- 219135
- Gene Name
- myotubularin related protein 6
- Gene Aliases
- 4022440C11Rik, AI428804, AU041072
- Chromosome Location
- Chr. 14: 60265183 - 60302371 on Build GRCm38
- UniGene ID
- Mm.247007
- Species
- Mus musculus
- Gene Symbol
- Nupl1
- Entrez Gene ID
- 71844
- Gene Name
- nucleoporin like 1
- Gene Aliases
- 1700017F11Rik, mKIAA0410
- Chromosome Location
- Chr. 14: 60205610 - 60251502 on Build GRCm38
- UniGene ID
- Mm.258051
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029688.1 |
- |
|
1871 |
|
View Full Product Details