Gene Transcripts

Gene Symbol
Top2a
Entrez Gene ID
21973
Gene Name
topoisomerase (DNA) II alpha
Gene Aliases
Top-2
Chromosome Location
Chr. 11: 98992947 - 99024206 on Build GRCm38
UniGene ID
Mm.4237
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011623.2 NP_035753.2 7 787
GenBank mRNA BC008170.1 - 7 775
BC057873.1 - 7 757
D12513.1 - 7 713

View Full Product Details