Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011692.2 | NP_035822.2 | 4 | 342 | |
GenBank mRNA | AK002282.1 | - | 4 | 312 | |
AK008847.1 | - | 4 | 312 | ||
AK077923.1 | - | 4 | 327 | ||
AK151527.1 | - | 4 | 312 | ||
AK153069.1 | - | 4 | 311 | ||
AK159284.1 | - | 4 | 312 | ||
AK167064.1 | - | 4 | 312 | ||
AK167742.1 | - | 4 | 343 | ||
AK169185.1 | - | 4 | 312 | ||
BC132169.1 | - | 4 | 285 | ||
BC132171.1 | - | 4 | 285 | ||
BY134471.1 | - | 4 | 335 | ||
DV066998.1 | - | 4 | 375 | ||
U96760.1 | - | 4 | 293 |