Gene Transcripts

Gene Symbol
Spidr
Entrez Gene ID
224008
Gene Name
scaffolding protein involved in DNA repair
Gene Aliases
2310008H04Rik, Kiaa0146
Chromosome Location
Chr. 16: 15889224 - 16146851 on Build GRCm38
UniGene ID
Mm.244703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146068.4 NP_666180.2 2 201
GenBank mRNA AK044836.1 - 2 184
AK052832.1 - 2 166
AK075871.1 - 2 166
AK146053.1 - 2 201
AK172902.1 - 2 154
BC089026.1 - 2 138 30615298

View Full Product Details