Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
226043
Gene Name
COBW domain containing 1
Gene Aliases
AV349248, BC018472
Chromosome Location
Chr. 19: 24919916 - 24961616 on Build GRCm38
UniGene ID
Mm.272127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146097.3 NP_666209.1 3, 4 424
GenBank mRNA AK132291.1 - 3, 4 410
BC018472.1 - 3, 4 366 3661842
BC053745.1 - 3, 4 459 6505520

View Full Product Details