Gene Transcripts

Gene Symbol
Epb4.1l5
Entrez Gene ID
226352
Gene Name
erythrocyte protein band 4.1 like 5
Gene Aliases
1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
Chromosome Location
Chr. 1: 119545032 - 119649136 on Build GRCm38
UniGene ID
Mm.253156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113416.1 NP_001106887.1 11, 12 1101
NM_145506.4 NP_663481.2 11, 12 1101
GenBank mRNA AK034676.1 - 11, 12 1023
AK037024.1 - 11, 12 1057
AK040354.1 - 11, 12 1069
AK044959.1 - 11, 12 1039
AK044996.1 - 11, 12 1047
AK047523.1 - 11, 12 1028
AK079301.1 - 11, 12 1052
AK136125.1 - 11, 12 1056
AK161141.1 - 11, 12 1052
AK167449.1 - 11, 12 1101
AK173198.1 - 11, 12 1035
BC003937.1 - 11, 12 1086 3486543
BC011476.1 - 11, 12 1035 4014009

View Full Product Details