Gene Transcripts

Gene Symbol
Zfp11
Entrez Gene ID
22648
Gene Name
zinc finger protein 11
Gene Aliases
6720465D24, Krox-6, Zfp-11
Chromosome Location
Chr. 5: 129654595 - 129670088 on Build GRCm38
UniGene ID
Mm.135067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172462.4 NP_766050.1 3 670
GenBank mRNA AK032872.1 - 3 685
AK085077.1 - 2 656
BC058553.1 - 3 668
BC094590.1 - 2 655 30616053
BC139312.1 - 2 650 8862334
BC139315.1 - 2 650 8862337

View Full Product Details