Gene Transcripts

Gene Symbol
Znhit6
Entrez Gene ID
229937
Gene Name
zinc finger, HIT type 6
Gene Aliases
2410019A14Rik
Chromosome Location
Chr. 3: 145576197 - 145605245 on Build GRCm38
UniGene ID
Mm.24586
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081094.1 NP_001074563.1 1, 2 799
GenBank mRNA AK148729.1 - 1, 2 776
AK167835.1 - 2, 3 221
AK168844.1 - 2, 3 215
BC147498.1 - 1, 2 685 9056360
BC150966.1 - 1, 2 685 9087878
BC150967.1 - 1, 2 685 9087879
BC151198.1 - 1, 2 685 9088086

View Full Product Details