Gene Transcripts

Gene Symbol
Usp1
Entrez Gene ID
230484
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
Chromosome Location
Chr. 4: 98923622 - 98935543 on Build GRCm38
UniGene ID
Mm.371692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301414.1 NP_001288343.1 6 1546
NM_146144.4 NP_666256.2 6 1527
GenBank mRNA AK080882.1 - 6 1528
BC018179.1 - 6 1407 3585191
BC020007.1 - 6 1379 3980469
BC023812.1 - 3 701

View Full Product Details