Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178257.2 | NP_839988.1 | 7 | 865 | |
GenBank mRNA | AK141289.1 | - | 7 | 866 | |
AY103454.1 | - | 7 | 804 | ||
BC140972.1 | - | 7 | 871 | 9055584 | |
BC140973.1 | - | 7 | 871 | 9055585 |