Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173413.3 | NP_775589.1 | 2 | 222 | |
GenBank mRNA | AB232601.1 | - | 2 | 150 | |
AK031251.1 | - | 2 | 169 | ||
AK051218.1 | - | 2 | 172 | ||
AK084650.1 | - | 2 | 172 | ||
AK155621.1 | - | 2 | 172 | ||
BC059208.1 | - | 2 | 204 | 6405462 | |
BC157974.1 | - | 2 | 170 | 9007296 | |
BY322741.1 | - | 2 | 223 |