Gene Transcripts

Gene Symbol
Mapk7
Entrez Gene ID
23939
Gene Name
mitogen-activated protein kinase 7
Gene Aliases
BMK-1, BMK1, ERK-5, ERK5, Erk5-T, PRKM7
Chromosome Location
Chr. 11: 61488812 - 61494266 on Build GRCm38
UniGene ID
Mm.38172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291033.1 NP_001277962.1 3, 4 699
NM_001291034.1 NP_001277963.1 2, 3 774
NM_001291035.1 NP_001277964.1 2, 3 915
NM_001291036.1 NP_001277965.1 3, 4 764
NM_001291037.1 NP_001277966.1 2, 3 1013
NM_011841.2 NP_035971.1 3, 4 649
GenBank mRNA AB019373.1 - 3, 4 599
AF126159.1 - 2, 3 412
AF126160.1 - 2, 3 462
AF126161.1 - 2, 3 832
AK148119.1 - 3, 4 631
AK155187.1 - 3, 4 567
AY534740.1 - 3, 4 583
BC013745.1 - 2, 3 932
BC033598.1 - 3, 4 739
BC051020.1 - 1, 2 1293
BC058096.1 - 1, 2 1293
BC082315.1 - 2, 3 836
BC100398.1 - 2, 3 716 30470655

View Full Product Details