Gene Transcripts

Gene Symbol
Cntnap5b
Entrez Gene ID
241175
Gene Name
contactin associated protein-like 5B
Gene Aliases
C230078M14Rik, Caspr5-2
Chromosome Location
Chr. 1: 99772247 - 100486066 on Build GRCm38
UniGene ID
Mm.251301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172851.2 NP_766439.2 11 1716
GenBank mRNA AK082656.1 - 5 774
BC145618.1 - 4 481

View Full Product Details