Gene Transcripts

Gene Symbol
Olfr1324
Entrez Gene ID
258289
Gene Name
olfactory receptor 1324
Gene Aliases
MOR128-2
Chromosome Location
Chr. X: 50425496 - 50426494 on Build GRCm38
UniGene ID
Mm.451653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146292.1 NP_666404.1 1 607
GenBank mRNA BC119382.1 - 1 695
BC120627.1 - 1 695 40129550

View Full Product Details