Gene Transcripts

Gene Symbol
Mok
Entrez Gene ID
26448
Gene Name
MOK protein kinase
Gene Aliases
RAGE1, Rage, Stk30
Chromosome Location
Chr. 12: 110807798 - 110840957 on Build GRCm38
UniGene ID
Mm.140948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011973.2 NP_036103.1 2 276
GenBank mRNA AB022695.1 - 2 276
BC049604.1 - 2 264

View Full Product Details