Gene Transcripts

Gene Symbol
Ppp2r5e
Entrez Gene ID
26932
Gene Name
protein phosphatase 2, regulatory subunit B', epsilon
Gene Aliases
4633401M22Rik, AI449017, B56beta
Chromosome Location
Chr. 12: 75449463 - 75596250 on Build GRCm38
UniGene ID
Mm.259626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012024.2 NP_036154.1 4 973
GenBank mRNA AK053228.1 - 3 823
AK088366.1 - 4 945
AK132432.1 - 4 1006
BC085149.1 - 4 973 6848033
BC094387.1 - 4 965
U49728.1 - 2 250

View Full Product Details