Gene Transcripts

Gene Symbol
Abca8b
Entrez Gene ID
27404
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8b
Gene Aliases
Abca8
Chromosome Location
Chr. 11: 109933406 - 109995846 on Build GRCm38
UniGene ID
Mm.341004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013851.2 NP_038879.2 3 369
GenBank mRNA AF498362.1 - 3 369
AK138196.1 - 3 370
AK173034.1 - 3 296
BC089557.1 - 3 217
BC141401.1 - 3 332 9056014

View Full Product Details