Gene Transcripts

Gene Symbol
Hip1r
Entrez Gene ID
29816
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
AA410023, mKIAA0655
Chromosome Location
Chr. 5: 123973607 - 124003215 on Build GRCm38
UniGene ID
Mm.149954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145070.3 NP_659507.3 5 515
GenBank mRNA AF221713.1 - 5 380
AK014542.1 - 5 493
AK129182.1 - 5 471
AK146663.1 - 5 396
BC062914.1 - 5 396
BC065101.1 - 5 504 30535741

View Full Product Details