Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289595.1 | NP_001276524.1 | 9 | 969 | |
NM_001289596.1 | NP_001276525.1 | 10 | 1022 | ||
NM_178766.5 | NP_848881.2 | 10 | 1041 | ||
GenBank mRNA | AK044260.1 | - | 1042 | ||
AK045863.1 | - | 10 | 1029 | ||
AK050815.1 | - | 2 | 2349 | ||
AK079538.1 | - | 10 | 971 | ||
AK084797.1 | - | 9 | 909 | ||
AK088444.1 | - | 10 | 1007 | ||
AK136770.1 | - | 10 | 1003 | ||
AK163037.1 | - | 10 | 2756 | ||
BC083103.1 | - | 9 | 819 | 30726859 |