Gene Transcripts

Gene Symbol
Snrnp200
Entrez Gene ID
320632
Gene Name
small nuclear ribonucleoprotein 200 (U5)
Gene Aliases
A330064G03Rik, Ascc3l1, BC011390, HELIC2, U5-200-KD, U5-200KD
Chromosome Location
Chr. 2: 127208404 - 127240451 on Build GRCm38
UniGene ID
Mm.215860
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177214.4 NP_796188.2 22 3053
GenBank mRNA AK173025.1 - 17 2282
BC063261.1 - 22 3053 6408607

View Full Product Details