Gene Transcripts

Gene Symbol
Usp44
Entrez Gene ID
327799
Gene Name
ubiquitin specific peptidase 44
Gene Aliases
E430004F17Rik
Chromosome Location
Chr. 10: 93819968 - 93860738 on Build GRCm38
UniGene ID
Mm.491598
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183199.3 NP_899022.2 4 1740
GenBank mRNA AK088109.1 - 4 1741
BC111886.1 - 2 1575 40060450

View Full Product Details