Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081201.1 | NP_001074670.1 | 12 | 1374 | |
GenBank mRNA | AK144417.1 | - | 12 | 1326 | |
AK165377.1 | - | 11 | 1185 | ||
AK172656.1 | - | 11 | 1250 | ||
BC147500.1 | - | 12 | 1330 | 9056363 | |
BC157932.1 | - | 12 | 1330 | 9088105 | |
BC157988.1 | - | 12 | 1330 | 9088143 |