Gene Transcripts

Gene Symbol
Gm1527
Entrez Gene ID
385263
Gene Name
predicted gene 1527
Gene Aliases
Gm1136
Chromosome Location
Chr. 3: 28892617 - 28926724 on Build GRCm38
UniGene ID
Mm.60893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033479.4 NP_001028651.1 8 865
GenBank mRNA AK132999.1 - 8 864

View Full Product Details