Gene Transcripts

Gene Symbol
Pnmal2
Entrez Gene ID
434128
Gene Name
PNMA-like 2
Gene Aliases
EG434128
Chromosome Location
Chr. 7: 16944682 - 16948828 on Build GRCm38
UniGene ID
Mm.483018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099636.2 NP_001093106.1 1 976
GenBank mRNA AK140282.1 - 1 977
AK143183.1 - 1 955

View Full Product Details