Gene Transcripts

Gene Symbol
Umodl1
Entrez Gene ID
52020
Gene Name
uromodulin-like 1
Gene Aliases
D17Ertd488e
Chromosome Location
Chr. 17: 30954640 - 31010710 on Build GRCm38
UniGene ID
Mm.25923
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177465.4 NP_803416.2 15 2956
GenBank mRNA AK040446.1 - 7 1457
AY771616.1 - 16 2873
AY771617.1 - 15 2960
AY771618.1 - 15 2615
BC051427.1 - 15 2611
BC132381.1 - 2 145

View Full Product Details