Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199044.1 | NP_001185973.1 | 19 | 5027 | |
NM_020027.3 | NP_064411.2 | 19 | 5027 | ||
GenBank mRNA | AK220526.1 | - | 18 | 4807 | |
BC006664.1 | - | 4 | 1197 | ||
BC034134.1 | - | 3 | 269 | ||
BC052630.1 | - | 19 | 4998 | ||
BC053522.1 | - | 19 | 5003 | 6491366 |