Gene Transcripts

Gene Symbol
Rabep1
Entrez Gene ID
54189
Gene Name
rabaptin, RAB GTPase binding effector protein 1
Gene Aliases
Rab5ep, rabaptin-5, rabaptin-5alpha
Chromosome Location
Chr. 11: 70844723 - 70943105 on Build GRCm38
UniGene ID
Mm.7087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291141.1 NP_001278070.1 10 1775
NM_001291142.1 NP_001278071.1 10 1784
NM_001291143.1 NP_001278072.1 12 1975
NM_019400.3 NP_062273.2 11 1904
GenBank mRNA AB001750.1 - 12 1811
AF248489.1 - 10 1570
AF248490.1 - 10 1561
AK134587.1 - 11 1905
BC003921.1 - 2 525
BC060166.1 - 11 1922
BC080274.1 - 11 1876
BC129853.1 - 11 1723 40109688
BC129854.1 - 11 1723 40109689
D86066.1 - 11 1740

View Full Product Details