Gene Transcripts

Gene Symbol
Paxip1
Entrez Gene ID
55982
Gene Name
PAX interacting (with transcription-activation domain) protein 1
Gene Aliases
D5Ertd149e, PTIP
Chromosome Location
Chr. 5: 27740080 - 27792079 on Build GRCm38
UniGene ID
Mm.277190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018878.3 NP_061366.2 18, 19 3281
GenBank mRNA AF104261.1 - 18, 19 3281
AK144606.1 - 18, 19 3143
AK148141.1 - 18, 19 3154
BC066014.1 - 18, 19 3070 5704566

View Full Product Details