Gene Transcripts

Gene Symbol
Dpt
Entrez Gene ID
56429
Gene Name
dermatopontin
Gene Aliases
1810032B19Rik, 5033416F05Rik, EQ-1, Eq1
Chromosome Location
Chr. 1: 164796732 - 164824266 on Build GRCm38
UniGene ID
Mm.28935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019759.2 NP_062733.1 2 408
GenBank mRNA AF143374.1 - 2 408
AK019890.1 - 2 497
BC046420.1 - 2 434 4165966

View Full Product Details