Gene Transcripts

Gene Symbol
Vps29
Entrez Gene ID
56433
Gene Name
vacuolar protein sorting 29 (S. pombe)
Gene Aliases
2010015D08Rik, AW049835, PEP11
Chromosome Location
Chr. 5: 122354377 - 122364981 on Build GRCm38
UniGene ID
Mm.216528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019780.1 NP_062754.1 4 551
GenBank mRNA AB578899.1 - 3 323
AB578900.1 - 3 323
AB578901.1 - 3 323
AF193794.1 - 4 551
AK004103.1 - 5 568
AK008248.1 - 5 571
AK036426.1 - 4 541
AK150330.1 - 4 561

View Full Product Details