Gene Transcripts

Gene Symbol
Olfr1509
Entrez Gene ID
57271
Gene Name
olfactory receptor 1509
Gene Aliases
MOR244-3, Mor83, Or83
Chromosome Location
Chr. 14: 52450393 - 52451402 on Build GRCm38
UniGene ID
Mm.88824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020514.2 NP_065260.2 1 540
GenBank mRNA BC119195.1 - 1 540
BC119199.1 - 1 540 8733948

View Full Product Details