Gene Transcripts

Gene Symbol
Nisch
Entrez Gene ID
64652
Gene Name
nischarin
Gene Aliases
1200007D05Rik, 3202002H23Rik, AW494485, I-1
Chromosome Location
Chr. 14: 31170928 - 31206826 on Build GRCm38
UniGene ID
Mm.298728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022656.2 NP_073147.2 11, 12 1475
GenBank mRNA AF315344.1 - 7, 8 910
AK004628.1 - 11, 12 1312
BC003270.1 - 11, 12 1337
BC066838.1 - 11, 12 1333
BC089329.1 - 6, 7 694
BC108364.1 - 6, 7 670
BC144980.1 - 7, 8 804

View Full Product Details