Gene Transcripts

Gene Symbol
Cuedc2
Entrez Gene ID
67116
Gene Name
CUE domain containing 2
Gene Aliases
3010002G01Rik
Chromosome Location
Chr. 19: 46329812 - 46338660 on Build GRCm38
UniGene ID
Mm.218848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164290.1 NP_001157762.1 4 296
NM_001164291.1 NP_001157763.1 3 188
NM_001164292.1 NP_001157764.1 3 185
NM_001164293.1 NP_001157765.1 3 188
NM_001164294.1 NP_001157766.1 3 188
NM_001164295.1 NP_001157767.1 3 185
NM_024192.2 NP_077154.2 4 287
GenBank mRNA AB232359.1 - 2 86
AK003457.1 - 3 143
AK008842.1 - 3 151
AK013884.1 - 3 146
AK207375.1 - 4 213
BC003442.1 - 4 251 2650719
BY279028.1 - 3 187
BY312150.1 - 3 149
CK625575.1 - 3 114
CX207621.1 - 3 487
CX218143.1 - 4 226
CX223880.1 - 4 251

View Full Product Details