Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026510.1 | NP_080786.1 | 3 | 555 | |
GenBank mRNA | AK009656.1 | - | 3 | 555 | |
AK017300.1 | - | 3 | 537 | ||
AK077929.1 | - | 3 | 536 | ||
AK145808.1 | - | 3 | 536 | ||
BC064776.1 | - | 2 | 523 | 30094986 |