Gene Transcripts

Gene Symbol
Sec62
Entrez Gene ID
69276
Gene Name
SEC62 homolog (S. cerevisiae)
Gene Aliases
3100002M17Rik, AI844545, AW545092, Dtrp1, HTP1, Tloc1
Chromosome Location
Chr. 3: 30792774 - 30821263 on Build GRCm38
UniGene ID
Mm.26017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027016.2 NP_081292.1 2 592
GenBank mRNA AK013922.1 - 2 407
AK088093.1 - 2 141
AK090078.1 - 2 142
BC017162.1 - 2 593
BC048532.1 - 2 142
BC053504.1 - 2 129
BC067202.1 - 2 127 6332701
BC071243.1 - 2 142

View Full Product Details