Gene Transcripts

Gene Symbol
Nmrk2
Entrez Gene ID
69564
Gene Name
nicotinamide riboside kinase 2
Gene Aliases
2310015C21Rik, Itgb1bp3, Mibp
Chromosome Location
Chr. 10: 81198170 - 81202212 on Build GRCm38
UniGene ID
Mm.437211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027120.2 NP_081396.1 5, 6 457
GenBank mRNA AK009352.1 - 5, 6 473
AK137483.1 - 5, 6 459
BC119244.1 - 5, 6 419 8733993
BC119246.1 - 5, 6 419 8733995

View Full Product Details