Gene Transcripts

Gene Symbol
Fam135b
Entrez Gene ID
70363
Gene Name
family with sequence similarity 135, member B
Gene Aliases
1700010C24Rik, A830008O07Rik
Chromosome Location
Chr. 15: 71438951 - 71728436 on Build GRCm38
UniGene ID
Mm.126450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177819.3 NP_808487.2 14 3503
GenBank mRNA AK005816.1 - 2 371
BC119174.1 - 2 119
BC137579.1 - 2 119

View Full Product Details